Diethylenetriaminepentaacetic dianhydride ≥98.0%

Supplier: TCI America

Synonyms: DTPA Dianhydride, DTPA anhydride, N,N-Bis[2-(2,6-dioxomorpholino)ethyl]glycine

D2424-5G D2424-25G
TCD2424-005GEA 206.66 CAD
TCD2424-005G TCD2424-025G
Diethylenetriaminepentaacetic dianhydride ≥98.0%
Diethylenetriaminepentaacetic dianhydride

CAS Number: 23911-26-4
MDL Number: MFCD00010697
Molecular Formula: C14H19N3O8
Molecular Weight: 357.32
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 182


Product Categories: Materials Chemistry, Monomers, Analytical Science, Chelating Agents for Magnetic Resonance Imaging (MRI), Complexones (Chelating Agents)
Other Product Names / Synonyms: N,N-Bis[2-(2,6-dioxomorpholino)ethyl]glycine, DTPA Dianhydride

Formula: C₁₄H₁₉N₃O₈
MW: 357.32 g/mol
Melting Pt: 182 °C
MDL Number: MFCD00010697
CAS Number: 23911-26-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce67dd2aa30c50a125b728b9053d1c9182fae433&vid=ce67dd2aa30c50a125b728b9053d1c9182fae433&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR