Dicyclohexylphenylphosphine ≥95.0%

Supplier: TCI America

D2411-1G D2411-5G
TCD2411-1GEA 120.54 CAD
TCD2411-1G TCD2411-005G
Dicyclohexylphenylphosphine ≥95.0%
Dicyclohexylphenylphosphine

CAS Number: 6476-37-5
MDL Number: MFCD00003854
Molecular Formula: C18H27P
Molecular Weight: 274.39
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 58


Product Categories: Synthetic Organic Chemistry, Cross-coupling Reaction using Transition Metal Catalysts, C-H Bond Activation Reaction, Mitsunobu Reaction, Ligands
Other Product Names / Synonyms: Phenylphosphinodicyclohexane

Formula: C₁₈H₂₇P
Melting Pt: 58 °C
MDL Number: MFCD00003854
CAS Number: 6476-37-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=73529719e81e9aa56eca1d97615413cef7908220&vid=73529719e81e9aa56eca1d97615413cef7908220&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR