2,4-Dichloro-5-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D2386-5G
D2386-25G
TCD2386-5GEA
65.85
CAD
TCD2386-5G
TCD2386-25G
2,4-Dichloro-5-fluorobenzoic acid ≥98.0% (by GC, titration analysis)
2,4-Dichloro-5-fluorobenzoic acid
CAS Number: 86522-89-6
MDL Number: MFCD00075281
Molecular Formula: C7H3Cl2FO2
Molecular Weight: 209.00
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 144
Formula:
C₇H₃Cl₂FO₂ Melting Pt: 144 °C |
MDL Number:
MFCD00075281 CAS Number: 86522-89-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...