2,6-Diacetylpyridine ≥98.0%
Supplier: TCI America
D2377-25G
D2377-5G
TCD2377-25GEA
654.59
CAD
TCD2377-25G
TCD2377-005G
2,6-Diacetylpyridine ≥98.0%
2,6-Diacetylpyridine
CAS Number: 1129-30-2
MDL Number: MFCD00006304
Molecular Formula: C9H9NO2
Molecular Weight: 163.18
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 126
Melting point (°C): 81
Formula:
C₉H₉NO₂ Boiling Pt: 126 °C Melting Pt: 81 °C |
MDL Number:
MFCD00006304 CAS Number: 1129-30-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...