3,4-Difluorobenzaldehyde ≥97.0%

Supplier: TCI America

D2340-5G D2340-25G
TCD2340-5GEA 85.32 CAD
TCD2340-5G TCD2340-25G
3,4-Difluorobenzaldehyde ≥97.0%
3,4-Difluorobenzaldehyde

CAS Number: 34036-07-2
MDL Number: MFCD00010328
Molecular Formula: C7H4F2O
Molecular Weight: 142.10
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 180
Flash Point (°C): 66
Specific Gravity (20/20): 1.31

Formula: C₇H₄F₂O
MW: 142.1 g/mol
Boiling Pt: 180 °C
Density: 1.31 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00010328
CAS Number: 34036-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=858223393f3344763cc5cfc1e3ed6c30f75bd803&vid=8019c4183ecf88bdf866a4c74c1624cf3ebc57aa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR