Diphenyliodonium trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D2253-1G D2253-5G D2253-25G
TCD2253-001GEA 134.3 CAD
TCD2253-001G TCD2253-005G TCD2253-25G
Diphenyliodonium trifluoromethanesulfonate ≥98.0% (by titrimetric analysis)
Diphenyliodonium trifluoromethanesulphonate

CAS Number: 66003-76-7
MDL Number: MFCD00191356
Molecular Formula: C13H10F3IO3S
Molecular Weight: 430.18
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 178

Formula: C₁₃H₁₀F₃IO₃S
Melting Pt: 178 °C
MDL Number: MFCD00191356
CAS Number: 66003-76-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5b5731ad65cf9da9c001537c274c94903ea34fd7&vid=5b5731ad65cf9da9c001537c274c94903ea34fd7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR