Ethyl diacetoacetate ≥96.0%
Supplier: TCI America
D2170-25G
D2170-5G
TCD2170-025GEA
145.36
CAD
TCD2170-025G
TCD2170-5G
Ethyl diacetoacetate ≥96.0%
Ethyl diacetoacetate
CAS Number: 603-69-0
MDL Number: MFCD00009864
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >96.0% (GC)
Form: Clear Liquid
Boiling point (°C): 103
Flash Point (°C): 85
Specific Gravity (20/20): 1.10
Other Product Names / Synonyms: Ethyl 2-Acetyl-3-oxobutyrate, Diacetoacetic Acid Ethyl Ester
Formula:
C₈H₁₂O₄ Boiling Pt: 103 °C Density: 1.10 (20 °C) Flash Pt: 85 °C |
MDL Number:
MFCD00009864 CAS Number: 603-69-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...