(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide ≥97.0%

Supplier: TCI America

Synonyms: ((1,3-Dioxolan-2-yl)methyl)triphenylphosphonium bromide

D2164-5G D2164-25G
TCD2164-005GEA 61.62 CAD
TCD2164-005G TCD2164-025G
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide ≥97.0%
(1,3-Dioxolan-2-ylmethyl)triphenylphosphonium bromide

CAS Number: 52509-14-5
MDL Number: MFCD00011966
Molecular Formula: C22H22BrO2P
Molecular Weight: 429.29
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 191

Formula: C₂₂H₂₂BrO₂P
MW: 429.30 g/mol
Melting Pt: 191 °C
MDL Number: MFCD00011966
CAS Number: 52509-14-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d8f9d8c5d7f51bf2bdc89e7ccafbfa110da7929d&vid=d8f9d8c5d7f51bf2bdc89e7ccafbfa110da7929d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR