(S)-(-)-2-Methyl-CBS-oxazaborolidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: (S)-5,5-Diphenyl-2-methyl-3,4-propano-1,3,2-oxazaborolidine, (S)-Me-CBS Catalyst, (S)-3,3-Diphenyl-1-methyltetrahydro-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole, (S)-3,3-Diphenyl-1-methylpyrrolidino[1,2-c]-1,3,2-oxazaborole

D2131-1G D2131-5G
TCD2131-001GEA 121.66 CAD
TCD2131-001G TCD2131-5G
(S)-(-)-2-Methyl-CBS-oxazaborolidine ≥98.0% (by titrimetric analysis)
(S)-(-)-2-Methyl-CBS-oxazaborolidine

CAS Number: 112022-81-8
MDL Number: MFCD00078439
Molecular Formula: C18H20BNO
Molecular Weight: 277.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Specific rotation [a]20/D: -68 deg (C=1, MeOH)


Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts
Other Product Names / Synonyms: (S)-3,3-Diphenyl-1-methyltetrahydro-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole, (S)-Me-CBS Catalyst

Formula: C₁₈H₂₀BNO
MW: 277.17 g/mol
MDL Number: MFCD00078439
CAS Number: 112022-81-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a4833bd5d78ce35c0c2a56673003c07d7b557063&vid=16220c27362b41408a42ac1db509d08782e7b1bb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR