(R)-(+)-2-Methyl-CBS-oxazaborolidine ≥97.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(R)-Tetrahydro-1-methyl-3,3-diphenyl-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole
CAS Number: 112022-83-0
MDL Number: MFCD00078440
Molecular Formula: C18H20BNO
Molecular Weight: 277.17
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Specific rotation [a]20/D: 68 deg (C=1, MeOH)
Product Categories: Synthetic Organic Chemistry, Asymmetric Organocatalysts
Other Product Names / Synonyms: (R)-3,3-Diphenyl-1-methyltetrahydro-1H,3H-pyrrolo[1,2-c][1,3,2]oxazaborole, (R)-Me-CBS Catalyst
Formula:
C₁₈H₂₀BNO MW: 277.17 g/mol |
MDL Number:
MFCD00078440 CAS Number: 112022-83-0 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...