4,4'-Oxybis(benzoic acid) ≥98.0%
Supplier: TCI America
Synonyms:
4,4′-Oxybis(benzoic acid)
D2115-25G
D2115-100G
TCD2115-025GEA
48.98
CAD
TCD2115-025G
TCD2115-100G
4,4'-Oxybis(benzoic acid) ≥98.0%
4,4'-Oxybis(benzoic acid)
CAS Number: 2215-89-6
MDL Number: MFCD00013988
Molecular Formula: C14H10O5
Molecular Weight: 258.23
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Product Categories: Materials Chemistry, Monomers, Diphenyl Ethers (for High-Performance Polymer Research)
Other Product Names / Synonyms: Diphenyl Ether 4,4'-Dicarboxylic Acid, 4,4'-Oxybisbenzoic Acid
Formula:
C₁₄H₁₀O₅ MW: 258.23 g/mol |
MDL Number:
MFCD00013988 CAS Number: 2215-89-6 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...