1,4-Thioxane 1,1-dioxide ≥98.0%
Supplier: TCI America
D2108-25G
D2108-5G
TCD2108-025GEA
192.76
CAD
TCD2108-025G
TCD2108-5G
1,4-Thioxane 1,1-dioxide ≥98.0%
1,4-Thioxane 1,1-dioxide
CAS Number: 107-61-9
MDL Number: MFCD00011683
Molecular Formula: C4H8O3S
Molecular Weight: 136.17
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Melting point (°C): 133
Other Product Names / Synonyms: 1,4-Oxathiane 4,4-Dioxide, 1,4-Thioxane 1,1-Dioxide
Formula:
C₄H₈O₃S Melting Pt: 133 °C |
MDL Number:
MFCD00011683 CAS Number: 107-61-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...