4'-(Diethylamino)acetophenone ≥98.0%
Supplier: TCI America
Synonyms:
1-(4-(Diethylamino)phenyl)ethanone
D2084-1G
D2084-5G
TCD2084-001GEA
134.3
CAD
TCD2084-001G
TCD2084-005G
4'-(Diethylamino)acetophenone ≥98.0%
4'-(Diethylamino)acetophenone
CAS Number: 5520-66-1
MDL Number: MFCD00043651
Molecular Formula: C12H17NO
Molecular Weight: 191.27
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 49
Storage Temperature: 0-10°C
Formula:
C₁₂H₁₇NO Melting Pt: 49 °C |
MDL Number:
MFCD00043651 CAS Number: 5520-66-1 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...