(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol ≥97.0%

Supplier: TCI America

D2024-5G D2024-25G
TCD2024-005GEA 39.5 CAD
TCD2024-005G TCD2024-025G
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol ≥97.0%
(1S,2S)-1,2-Bis((R)-2,2-dimethyl-1,3-dioxolan-4-yl)ethane-1,2-diol

CAS Number: 1707-77-3
MDL Number: MFCD00003211
Molecular Formula: C12H22O6
Molecular Weight: 262.30
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 120
Specific rotation [a]20/D: 7 deg (C=5, CHCl3)

Formula: C₁₂H₂₂O₆
Melting Pt: 120 °C
MDL Number: MFCD00003211
CAS Number: 1707-77-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=69928a4d45395e928779cb2722add176d5ade406&vid=69928a4d45395e928779cb2722add176d5ade406&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR