1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose ≥97.0%

Supplier: TCI America

D1949-10G D1949-25G
TCD1949-010GEA 25.72 CAD
TCD1949-010G TCD1949-025G
1,2:5,6-Di-O-isopropylidene-α-D-glucofuranose ≥97.0%
1,2:5,6-Di-O-isopropylidene-alpha-D-glucofuranose

CAS Number: 582-52-5
MDL Number: MFCD00005544
Molecular Formula: C12H20O6
Molecular Weight: 260.29
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 110
Specific rotation [a]20/D: -18.5 deg (C=5, H2O)


Product Categories: Synthetic Organic Chemistry, Glycosylation, Chiral Building Blocks, Bioscience, Glucose, O-Substituted Sugars
Other Product Names / Synonyms: Diacetone-D-glucose

Formula: C₁₂H₂₀O₆
Melting Pt: 110 °C
MDL Number: MFCD00005544
CAS Number: 582-52-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6f4ce0149082f5c7e5ef6f32adb317755d4a6ae&vid=e5eceb3c308fd7ebbc50de7115bd0273521205de&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR