3-Decen-2-one ≥93.0%
Supplier: TCI America
D1933-25ML
D1933-5ML
TCD1933-025MLEA
101.12
CAD
TCD1933-025ML
TCD1933-5ML
3-Decen-2-one ≥93.0%
3-Decen-2-one
CAS Number: 10519-33-2
MDL Number: MFCD00015700
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >93.0% (GC)
Form: Clear Liquid
Boiling point (°C): 109
Flash Point (°C): 92
Specific Gravity (20/20): 0.85
Formula:
C₁₀H₁₈O Boiling Pt: 109 °C Density: 0.85 (20 °C) Flash Pt: 92 °C |
MDL Number:
MFCD00015700 CAS Number: 10519-33-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...