2,5-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
Supplier: TCI America
D1894-25G
D1894-5G
TCD1894-25GEA
189.6
CAD
TCD1894-25G
TCD1894-5G
2,5-Dimethoxybenzoic acid ≥98.0% (by GC, titration analysis)
2,5-Dimethoxybenzoic acid
CAS Number: 2785-98-0
MDL Number: MFCD00002436
Molecular Formula: C9H10O4
Molecular Weight: 182.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 78
Formula:
C₉H₁₀O₄ Melting Pt: 78 °C |
MDL Number:
MFCD00002436 CAS Number: 2785-98-0 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...