Ethyl-4,4-imethyl-3-oxovalerate ≥98.0%

Supplier: TCI America

D1891-5ML D1891-25ML
TCD1891-5MLEA 64.78 CAD
TCD1891-5ML TCD1891-025ML
Ethyl-4,4-imethyl-3-oxovalerate ≥98.0%
Ethyl-4,4-imethyl-3-oxovalerate

CAS Number: 17094-34-7
MDL Number: MFCD00042886
Molecular Formula: C9H16O3
Molecular Weight: 172.22
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 88
Flash Point (°C): 78
Specific Gravity (20/20): 0.97


Product Categories: Synthetic Organic Chemistry, ƒÀ-Keto Esters (Multi-component Reaction)
Other Product Names / Synonyms: 4,4-Dimethyl-3-oxovaleric Acid Ethyl Ester, Ethyl Pivaloylacetate, Pivaloylacetic Acid Ethyl Ester

Formula: C₉H₁₆O₃
Boiling Pt: 88 °C
Density: 0.97 (20 °C)
Flash Pt: 78 °C
MDL Number: MFCD00042886
CAS Number: 17094-34-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d475cf7b58e684297ee448ec76622ff55eb3cdaa&vid=d475cf7b58e684297ee448ec76622ff55eb3cdaa&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR