1,3-Dichloro-5-iodobenzene ≥98.0%

Supplier: TCI America

D1866-5G D1866-25G
TCD1866-5GEA 69.14 CAD
TCD1866-5G TCD1866-25G
1,3-Dichloro-5-iodobenzene ≥98.0%
1,3-Dichloro-5-iodobenzene

CAS Number: 3032-81-3
MDL Number: MFCD00001047
Molecular Formula: C6H3Cl2I
Molecular Weight: 272.89
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 127
Melting point (°C): 58


Other Product Names / Synonyms: 3,5-Dichloroiodobenzene

Formula: C₆H₃Cl₂I
Boiling Pt: 127 °C
Melting Pt: 58 °C
MDL Number: MFCD00001047
CAS Number: 3032-81-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1b8306e33a4b9bd1497c02ead1db679dd72aab6&vid=28d1fd9656292e94168e7f255ee083e1456f2d35&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR