2,4-Dimethoxybenzyl alcohol ≥98.0%

Supplier: TCI America

D1835-25G D1835-5G
TCD1835-25GEA 195.92 CAD
TCD1835-25G TCD1835-5G
2,4-Dimethoxybenzyl alcohol ≥98.0%
2,4-Dimethoxybenzyl alcohol

CAS Number: 7314-44-5
MDL Number: MFCD00004614
Molecular Formula: C9H12O3
Molecular Weight: 168.19
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 36
Flash Point (°C): 113

Formula: C₉H₁₂O₃
MW: 168.19
Boiling Pt: 135 °C
Melting Pt: 36 °C
Flash Pt: 113 °C
MDL Number: MFCD00004614
CAS Number: 7314-44-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=16a62edb74c3d8e6af170f7b8a103e2b7d960e22&vid=16a62edb74c3d8e6af170f7b8a103e2b7d960e22&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR