Di(N-succinimidyl) carbonate ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
N-Succinimidyl Carbonate, N,N'-Disuccinimidyl Carbonate, DSC, Di-(N,N'-succinimidyl) carbonate, Bis(2,5-dioxopyrrolidin-1-yl) carbonate
CAS Number: 74124-79-1
MDL Number: MFCD00009767
Molecular Formula: C9H8N2O7
Molecular Weight: 256.17
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 215
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Condensation & Active Esterification, Phosgene & Thiophosgene Alternatives, Peptide Synthesis, Bioscience, Crosslinkers
Other Product Names / Synonyms: N-Succinimidyl Carbonate, N,N'-Disuccinimidyl Carbonate, DSC
Formula:
C₉H₈N₂O₇ MW: 256.17 g/mol Melting Pt: 215 °C |
MDL Number:
MFCD00009767 CAS Number: 74124-79-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...