[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphonium bromide ≥98.0%

Supplier: TCI America

Synonyms: (2-(1,3-Dioxan-2-yl)ethyl)triphenylphosphonium bromide

D1655-25G D1655-5G
TCD1655-25GEA 516.66 CAD
TCD1655-25G TCD1655-005G
[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphonium bromide ≥98.0%
[2-(1,3-Dioxan-2-yl)ethyl]triphenylphosphonium bromide

CAS Number: 69891-92-5
MDL Number: MFCD00012000
Molecular Formula: C24H26BrO2P
Molecular Weight: 457.35
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 189

Formula: C₂₄H₂₆BrO₂P
MW: 457.36 g/mol
Melting Pt: 189 °C
MDL Number: MFCD00012000
CAS Number: 69891-92-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=efc8f2589d4d9ca07ffb0156df8208e88ba9f829&vid=efc8f2589d4d9ca07ffb0156df8208e88ba9f829&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR