N,N-Dimethylmethyleneammonium iodide ≥97.0% (by titrimetric analysis)
Supplier: TCI America
D1646-25G
D1646-5G
TCD1646-025GEA
303.36
CAD
TCD1646-025G
TCD1646-005G
N,N-Dimethylmethyleneammonium iodide ≥97.0% (by titrimetric analysis)
N,N-Dimethylmethyleneammonium iodide
CAS Number: 33797-51-2
MDL Number: MFCD00011810
Molecular Formula: C3H8IN
Molecular Weight: 185.01
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Other Product Names / Synonyms: Eschenmoser's Salt, N,N-Dimethylmethyleneiminium Iodide
Formula:
C₃H₈IN |
MDL Number:
MFCD00011810 CAS Number: 33797-51-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...