(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol ≥98.0% (by HPLC)

Supplier: TCI America

D1623-5G D1623-1G
TCD1623-5GEA 252.96 CAD
TCD1623-5G TCD1623-001G
(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol ≥98.0% (by HPLC)
(-)-1,4-Di-O-tosyl-2,3-O-isopropylidene-L-threitol

CAS Number: 37002-45-2
MDL Number: MFCD00003212
Molecular Formula: C21H26O8S2
Molecular Weight: 470.55
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 93
Specific rotation [a]20/D: -23 deg (C=8.8, CH3CN)


Product Categories: Synthetic Organic Chemistry, Sulfonic Acid Derivatives, Chiral Building Blocks, Bioscience, Sugar Alcohols, Sugars (Others), O-Substituted Sugars
Other Product Names / Synonyms: (4S,5S)-(-)-4,5-Bis(tosyloxymethyl)-2,2-dimethyl-1,3-dioxolane, (4S,5S)-(-)-2,2-Dimethyl-4,5-bis(tosyloxymethyl)-1,3-dioxide, (4S,5S)-(-)-2,3-O-Isopropylidene-1,4-di-O-tosyl-L-threitol

Formula: C₂₁H₂₆O₈S₂
Melting Pt: 93 °C
MDL Number: MFCD00003212
CAS Number: 37002-45-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7e0cae9f20683dc19ad1fce9d47c48dd88291f89&vid=7e0cae9f20683dc19ad1fce9d47c48dd88291f89&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR