2,3-Dihydrobenzo[b]furan ≥98.0%

Supplier: TCI America

D1583-25G D1583-5G
TCD1583-25GEA 204.93 CAD
TCD1583-25G TCD1583-5G
2,3-Dihydrobenzo[b]furan ≥98.0%
2,3-Dihydrobenzo[b]furan

CAS Number: 496-16-2
MDL Number: MFCD00005855
Molecular Formula: C8H8O
Molecular Weight: 120.15
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 189
Flash Point (°C): 66
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: Coumaran

Formula: C₈H₈O
MW: 120.15 g/mol
Boiling Pt: 189 °C
Density: 1.09 (20 °C)
Flash Pt: 66 °C
MDL Number: MFCD00005855
CAS Number: 496-16-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=74fd5b8fcf6853454bf981e3272f29fa29f150e4&vid=74fd5b8fcf6853454bf981e3272f29fa29f150e4&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR