4,6-Dimethyl-2-mercaptopyrimidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

D1580-250G D1580-25G
TCD1580-250GEA 456.62 CAD
TCD1580-250G TCD1580-025G
4,6-Dimethyl-2-mercaptopyrimidine ≥98.0% (by titrimetric analysis)

CAS Number: 22325-27-5
MDL Number: MFCD00006079
Molecular Formula: C6H8N2S
Molecular Weight: 140.20
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 219


Other Product Names / Synonyms: 4,6-Dimethylpyrimidine-2-thiol, 2-Mercapto-4,6-dimethylpyrimidine

Formula: C₆H₈N₂S
Melting Pt: 219 °C
MDL Number: MFCD00006079
CAS Number: 22325-27-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b99c1fff06fc38d922ed556da0f1f3423f76012a&vid=d7d811688511e2c6c44bf4292c21a05a1cbf7c9d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR