1,3-Diphenylisobenzofuran ≥97.0%

Supplier: TCI America

D1520-25G D1520-5G
TCD1520-25GEA 440.82 CAD
TCD1520-25G TCD1520-005G
1,3-Diphenylisobenzofuran ≥97.0%
1,3-Diphenylisobenzofuran

CAS Number: 5471-63-6
MDL Number: MFCD00005931
Molecular Formula: C20H14O
Molecular Weight: 270.33
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Melting point (°C): 131

Formula: C₂₀H₁₄O
Melting Pt: 131 °C
MDL Number: MFCD00005931
CAS Number: 5471-63-6

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR