Dodecylamine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: Dodecan-1-amine hydrochloride, Laurylamine Hydrochloride

D1452-25G D1452-500G D1452-100G
TCD1452-025GEA 86.9 CAD
TCD1452-025G TCD1452-500G TCD1452-100G
Dodecylamine hydrochloride ≥98.0%
Dodecylamine hydrochloride

CAS Number: 929-73-7
MDL Number: MFCD00042017
Molecular Formula: C12H27N
Molecular Weight: 221.81
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Cationic Surfactants, Derivatizing Reagents for Solubility Improvement
Other Product Names / Synonyms: Laurylamine Hydrochloride

Formula: C₁₂H₂₇N
MDL Number: MFCD00042017
CAS Number: 929-73-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=653440ece023e46ff8ca24a4fb5edcec3d183bb3&vid=653440ece023e46ff8ca24a4fb5edcec3d183bb3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR