cis-1,4-Diacetoxy-2-butene ≥95.0%

Supplier: TCI America

D1358-25ML D1358-500ML
TCD1358-025MLEA 121.66 CAD
TCD1358-025ML TCD1358-500ML
cis-1,4-Diacetoxy-2-butene ≥95.0%
cis-1,4-Diacetoxy-2-butene

CAS Number: 25260-60-0
MDL Number: MFCD00059339
Molecular Formula: C8H12O4
Molecular Weight: 172.18
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Specific Gravity (20/20): 1.09


Other Product Names / Synonyms: cis-2-Butene-1,4-diol Diacetate

Formula: C₈H₁₂O₄
Boiling Pt: 110 °C
Density: 1.09 (20 °C)
MDL Number: MFCD00059339
CAS Number: 25260-60-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=729ea3ec86f8aa3bf825d2a38e7c42908ac58265&vid=729ea3ec86f8aa3bf825d2a38e7c42908ac58265&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR