1,4-Dithioerythritol ≥98.0%

Supplier: TCI America

Synonyms: Cleland's Reagent, (R*,S*)-1,4-Dimercaptobutane-2,3-diol, DTE, Erythro-1,4-Dimercaptobutane-2,3-diol, trans-1,4-Dimercaptobutane-2,3-diol, Dithioerythritol, DL-Dithioerythritol

D1320-25G D1320-5G
TCD1320-25GEA 777.36 CAD
TCD1320-25G TCD1320-5G
1,4-Dithioerythritol ≥98.0%
1,4-Dithioerythritol

CAS Number: 6892-68-8
MDL Number: MFCD00063750
Molecular Formula: C4H10O2S2
Molecular Weight: 154.24
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 84
Storage Temperature: 0-10°C


Product Categories: Materials Chemistry, Monomers, Bioscience, Sugar Alcohols, Erythroses, O-Substituted Sugars, Antioxidants
Other Product Names / Synonyms: 2,3-Dihydroxy-1,4-butanedithiol, 1,4-Dimercapto-2,3-butanediol, DTE

Formula: C₄H₁₀O₂S₂
MW: 154.25 g/mol
Boiling Pt: 286
Melting Pt: 84 °C
MDL Number: MFCD00063750
CAS Number: 6892-68-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=35a9d2b070163d4eb658520aac57758d19d4664d&vid=35a9d2b070163d4eb658520aac57758d19d4664d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR