2,5-Dithiobiurea ≥95.0%
Supplier: TCI America
Synonyms:
Hydrazine-1,2-bis(carbothioamide)
D1308-25G
D1308-10G
TCD1308-25GEA
211.72
CAD
TCD1308-25G
TCD1308-010G
2,5-Dithiobiurea ≥95.0%
2,5-Dithiobiurea
CAS Number: 142-46-1
MDL Number: MFCD00014471
Molecular Formula: C2H6N4S2
Molecular Weight: 150.22
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Melting point (°C): 196
Formula:
C₂H₆N₄S₂ MW: 150.22 g/mol Melting Pt: 196 °C |
MDL Number:
MFCD00014471 CAS Number: 142-46-1 |
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...