3-Methoxy-N,N-dimethylaniline ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D1288-5ML D1288-25ML
TCD1288-5MLEA 237 CAD
TCD1288-5ML TCD1288-025ML
3-Methoxy-N,N-dimethylaniline ≥97.0% (by GC, titration analysis)
3-Methoxy-N,N-dimethylaniline

CAS Number: 15799-79-8
MDL Number: MFCD00051779
Molecular Formula: C9H13NO
Molecular Weight: 151.21
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 123
Specific Gravity (20/20): 1.03


Other Product Names / Synonyms: 3-Dimethylaminoanisole, 3-Methoxy-N,N-dimethylaniline

Formula: C₉H₁₃NO
MW: 151.21 g/mol
Boiling Pt: 123 °C
Density: 1.03 (20 °C)
MDL Number: MFCD00051779
CAS Number: 15799-79-8
UN: 2810
ADR: 6.1 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c92fb2ce505ed3e86297c5f15a388622c5082ec9&vid=c92fb2ce505ed3e86297c5f15a388622c5082ec9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR