Diphenolic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 4,4-Bis(4-hydroxyphenyl)valeric Acid

D1274-500G D1274-25G
TCD1274-500GEA 739.44 CAD
TCD1274-500G TCD1274-025G
Diphenolic acid ≥98.0% (by GC, titration analysis)
Diphenolic acid

CAS Number: 126-00-1
MDL Number: MFCD00002800
Molecular Formula: C17H18O4
Molecular Weight: 286.33
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 177
Flash Point (°C): 208


Other Product Names / Synonyms: 4,4-Bis(4-hydroxyphenyl)valeric Acid

Formula: C₁₇H₁₈O₄
MW: 286.33 g/mol
Melting Pt: 177 °C
Flash Pt: 208 °C
MDL Number: MFCD00002800
CAS Number: 126-00-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=af17f91278ed68ff4c871a690a4b7528dbc489c3&vid=af17f91278ed68ff4c871a690a4b7528dbc489c3&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR