2-Decyn-1-ol ≥95.0%

Supplier: TCI America

D1264-5ML D1264-25ML
TCD1264-005MLEA 284.49 CAD
TCD1264-005ML TCD1264-25ML
2-Decyn-1-ol ≥95.0%
2-Decyn-1-ol

CAS Number: 4117-14-0
MDL Number: MFCD00041567
Molecular Formula: C10H18O
Molecular Weight: 154.25
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 113
Flash Point (°C): 110
Specific Gravity (20/20): 0.87


Other Product Names / Synonyms: Heptyl(hydroxymethyl)acetylene

Formula: C₁₀H₁₈O
Boiling Pt: 113 °C
Density: 0.87 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00041567
CAS Number: 4117-14-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c947d7850c4de0e6f3af90ee0cab1c009bacfc3b&vid=c947d7850c4de0e6f3af90ee0cab1c009bacfc3b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR