DL-2,4-Diaminobutyric acid dihydrochloride ≥98.0%
Supplier: TCI America
Synonyms:
H-DL-Dab·2HCl, H-DL-Dab.2HCl
D1175-25G
D1175-1G
D1175-5G
TCD1175-25GEA
2047.68
CAD
TCD1175-25G
TCD1175-001G
TCD1175-005G
DL-2,4-Diaminobutyric acid dihydrochloride ≥98.0%
DL-2,4-Diaminobutyric acid dihydrochloride
CAS Number: 65427-54-5
MDL Number: MFCD00012910
Molecular Formula: C4H10N2O2
Molecular Weight: 191.05
Purity/Analysis Method: >98.0% (N,T)
Form: Crystal
Melting point (°C): 203
Formula:
C₄H₁₀N₂O₂·₂HCl MW: 191.1 g/mol Melting Pt: 203 °C |
MDL Number:
MFCD00012910 CAS Number: 65427-54-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...