3,6-Dimethyl-4-octyne-3,6-diol ≥98.0%

Supplier: TCI America

D1168-25G D1168-500G
TCD1168-025GEA 47.4 CAD
TCD1168-025G TCD1168-500G
3,6-Dimethyl-4-octyne-3,6-diol ≥98.0%
3,6-Dimethyl-4-octyne-3,6-diol

CAS Number: 78-66-0
MDL Number: MFCD00004480
Molecular Formula: C10H18O2
Molecular Weight: 170.25
Purity/Analysis Method: >98.0% (GC)
Form: Crystal
Boiling point (°C): 135
Melting point (°C): 54
Flash Point (°C): 113


Other Product Names / Synonyms: 3,6-Dihydroxy-3,6-dimethyl-4-octyne

Formula: C₁₀H₁₈O₂
Boiling Pt: 135 °C
Melting Pt: 54 °C
Flash Pt: 113 °C
MDL Number: MFCD00004480
CAS Number: 78-66-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0943239385fd9382b5d4878ded6554840a40f7a1&vid=0943239385fd9382b5d4878ded6554840a40f7a1&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR