Bis(2-benzamidophenyl)disulfide ≥98.0% (by total nitrogen basis)

Supplier: TCI America

Synonyms: 2,2'-Dithiodibenzanilide, 2,2'-Dibenzamidodiphenyl Disulphide, Bis(2-benzamidophenyl) Disulfide, 2,2'-Dibenzamidodiphenyl Disulfide, Bis(2-benzamidophenyl) Disulphide

D1102-25G D1102-500G
TCD1102-025GEA 26.86 CAD
TCD1102-025G TCD1102-500G
Bis(2-benzamidophenyl)disulfide ≥98.0% (by total nitrogen basis)
Bis(2-benzamidophenyl)disulfide

CAS Number: 135-57-9
MDL Number: MFCD00043806
Molecular Formula: C26H20N2O2S2
Molecular Weight: 456.58
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 143
Flash Point (°C): 268


Other Product Names / Synonyms: 2,2'-Dibenzamidodiphenyl Disulfide, 2,2'-Dithiodibenzanilide

Formula: C₂₆H₂₀N₂O₂S₂
MW: 456.58 g/mol
Melting Pt: 143 °C
Flash Pt: 268 °C
MDL Number: MFCD00043806
CAS Number: 135-57-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fda7f00d627d80a24e6ef9f2c3dda62f62f0a3d2&vid=fda7f00d627d80a24e6ef9f2c3dda62f62f0a3d2&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR