1,4-Bis(2-hydroxyethoxy)benzene ≥95.0%

Supplier: TCI America

D1065-500G D1065-25G
TCD1065-500GEA 120.08 CAD
TCD1065-500G TCD1065-025G
1,4-Bis(2-hydroxyethoxy)benzene ≥95.0%
1,4-Bis(2-hydroxyethoxy)benzene

CAS Number: 104-38-1
MDL Number: MFCD00002861
Molecular Formula: C10H14O4
Molecular Weight: 198.22
Purity/Analysis Method: >95.0% (GC)
Form: Crystal
Melting point (°C): 105
Flash Point (°C): 212


Other Product Names / Synonyms: 1,4-Di(2-hydroxyethoxy)benzene, Hydroquinone Bis(2-hydroxyethyl) Ether, Hydroquinone Di(2-hydroxyethyl) Ether

Formula: C₁₀H₁₄O₄
Melting Pt: 105 °C
Flash Pt: 212 °C
MDL Number: MFCD00002861
CAS Number: 104-38-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=297844871d9024b8cba9d84bb34d070e72d3fdeb&vid=297844871d9024b8cba9d84bb34d070e72d3fdeb&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR