Diethylphenylphosphine ≥97.0% (by GC, titration analysis)

Supplier: TCI America

D1019-1ML D1019-5ML
TCD1019-1MLEA 235.42 CAD
TCD1019-1ML TCD1019-005ML
Diethylphenylphosphine ≥97.0% (by GC, titration analysis)
Diethylphenylphosphine

CAS Number: 1605-53-4
MDL Number: MFCD00015172
Molecular Formula: C10H15P
Molecular Weight: 166.20
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 220
Flash Point (°C): 85
Specific Gravity (20/20): 0.95

Formula: C₁₀H₁₅P
Boiling Pt: 220 °C
Density: 0.95 (20 °C)
Flash Pt: 85 °C
MDL Number: MFCD00015172
CAS Number: 1605-53-4
UN: 3278
ADR: 6.1,I
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b29c846a495d7f9f2952aecdba838be7916a72a9&vid=b29c846a495d7f9f2952aecdba838be7916a72a9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR