Erucic acid ≥85.0%
Supplier: TCI America
Synonyms:
(Z)-Docos-13-enoic acid
D0965-25G
D0965-100G
TCD0965-025GEA
37.92
CAD
TCD0965-025G
TCD0965-100G
Erucic acid ≥85.0%
Erucic acid
CAS Number: 112-86-7
MDL Number: MFCD00063188
Molecular Formula: C22H42O2
Molecular Weight: 338.58
Purity/Analysis Method: >85.0% (GC)
Form: Crystal
Flash Point (°C): 110
Freezing point (°C): 30
Storage Temperature: 0-10°C
Product Categories: Bioscience, Lipids, Higher Fatty Acids & Higher Alcohols
Other Product Names / Synonyms: cis-13-Docosenoic Acid
Formula:
C₂₂H₄₂O₂ Flash Pt: 110 °C |
MDL Number:
MFCD00063188 CAS Number: 112-86-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...