1,3-Di-o-tolylguanidine ≥99.0%

Supplier: TCI America

D0953-500G D0953-25G
TCD0953-500GEA 259.12 CAD
TCD0953-500G TCD0953-025G
1,3-Di-o-tolylguanidine ≥99.0%
1,3-Di-o-tolylguanidine

CAS Number: 97-39-2
MDL Number: MFCD00008513
Molecular Formula: C15H17N3
Molecular Weight: 239.32
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 175
Flash Point (°C): 170

Formula: C₁₅H₁₇N₃
Melting Pt: 175 °C
Flash Pt: 170 °C
MDL Number: MFCD00008513
CAS Number: 97-39-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=1bb011b3796d510a3ebd35eb3e1c0ec0b5bcc75f&vid=1bb011b3796d510a3ebd35eb3e1c0ec0b5bcc75f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 99.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR