1,3-Di(4-pyridyl)propane ≥97.0%

Supplier: TCI America

Synonyms: 4,4'-Trimethylenedipyridine

D0938-25G D0938-500G
TCD0938-025GEA 81.94 CAD
TCD0938-025G TCD0938-500G
1,3-Di(4-pyridyl)propane ≥97.0%
1,3-Di(4-pyridyl)propane

CAS Number: 17252-51-6
MDL Number: MFCD00038046
Molecular Formula: C13H14N2
Molecular Weight: 198.27
Purity/Analysis Method: >97.0% (GC)
Form: Crystal
Boiling point (°C): 162
Melting point (°C): 62
Flash Point (°C): 163

Formula: C₁₃H₁₄N₂
MW: 198.27 g/mol
Boiling Pt: 162 °C
Melting Pt: 62 °C
Flash Pt: 163 °C
MDL Number: MFCD00038046
CAS Number: 17252-51-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a38378f7cc8ae967a131607a76fa454abcd1e14d&vid=a38378f7cc8ae967a131607a76fa454abcd1e14d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR