Dicinnamalacetone ≥98.0% (by HPLC)

Supplier: TCI America

D0900-10G D0900-25G
TCD0900-010GEA 90.06 CAD
TCD0900-010G TCD0900-025G
Dicinnamalacetone ≥98.0% (by HPLC)
Dicinnamalacetone

CAS Number: 622-21-9
MDL Number: MFCD00004792
Molecular Formula: C21H18O
Molecular Weight: 286.37
Purity/Analysis Method: >98.0% (HPLC)
Form: Crystal
Melting point (°C): 143


Other Product Names / Synonyms: 1,9-Diphenyl-1,3,6,8-nonatetraen-5-one

Formula: C₂₁H₁₈O
Melting Pt: 143 °C
MDL Number: MFCD00004792
CAS Number: 622-21-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0424325d71831c62e4969d3a4b74ef377d0195c6&vid=0424325d71831c62e4969d3a4b74ef377d0195c6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (HPLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR