2,2'-Dinaphthyl ether ≥99.0%
Supplier: TCI America
D0811-10G
D0811-25G
TCD0811-010GEA
268.6
CAD
TCD0811-010G
TCD0811-025G
2,2'-Dinaphthyl ether ≥99.0%
2,2'-Dinaphthyl ether
CAS Number: 613-80-9
MDL Number: MFCD00015447
Molecular Formula: C20H14O
Molecular Weight: 270.33
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Boiling point (°C): 250
Melting point (°C): 105
Formula:
C₂₀H₁₄O Boiling Pt: 250 °C Melting Pt: 105 °C |
MDL Number:
MFCD00015447 CAS Number: 613-80-9 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...