Nordihydroguaiaretic acid ≥97.0% (by HPLC)

Supplier: TCI America

D0800-1G D0800-5G
TCD0800-001GEA 176.87 CAD
TCD0800-001G TCD0800-005G
Nordihydroguaiaretic acid ≥97.0% (by HPLC)
Nordihydroguaiaretic acid

CAS Number: 500-38-9
MDL Number: MFCD00002206
Molecular Formula: C18H22O4
Molecular Weight: 302.37
Purity/Analysis Method: >97.0% (HPLC)
Form: Crystal
Melting point (°C): 185


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Antioxidants, Phenylpropanoids, Polyphenols (Phytochemicals)
Other Product Names / Synonyms: NDGA, 4,4'-(2,3-Dimethyltetramethylene)dipyrocatechol

Formula: C₁₈H₂₂O₄
MW: 302.37 g/mol
Melting Pt: 185 °C
MDL Number: MFCD00002206
CAS Number: 500-38-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f65e61389007f9372d1cbb58066b870d918b7f6&vid=3f65e61389007f9372d1cbb58066b870d918b7f6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 97.0% (HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR