1,3-Bis(hydroxymethyl)urea ≥98.0% (by total nitrogen basis)
Supplier: TCI America
Synonyms:
N,N'-Bis(hydroxymethyl)urea, N,N'-Dimethylolurea
D0767-25G
D0767-500G
TCD0767-025GEA
39.5
CAD
TCD0767-025G
TCD0767-500G
1,3-Bis(hydroxymethyl)urea ≥98.0% (by total nitrogen basis)
1,3-Bis(hydroxymethyl)urea
CAS Number: 140-95-4
MDL Number: MFCD00014414
Molecular Formula: C3H8N2O3
Molecular Weight: 120.11
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 133
Storage Temperature: 0-10°C
Product Categories: Synthetic Organic Chemistry, Ureas (Multi-component Reaction)
Other Product Names / Synonyms: N,N'-Dimethylolurea
Formula:
C₃H₈N₂O₃ MW: 120.11 g/mol Melting Pt: 133 °C |
MDL Number:
MFCD00014414 CAS Number: 140-95-4 |
Specification Test Results
Purity/Analysis Method |
|
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...