4,4'-Dimethoxydiphenylamine ≥98.0%
Supplier: TCI America
Synonyms:
Bis(4-methoxyphenyl)amine
D0632-1G
D0632-5G
TCD0632-001GEA
61.07
CAD
TCD0632-001G
TCD0632-5G
4,4'-Dimethoxydiphenylamine ≥98.0%
4,4'-Dimethoxydiphenylamine
CAS Number: 101-70-2
MDL Number: MFCD00014895
Molecular Formula: C14H15NO2
Molecular Weight: 229.28
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 103
Product Categories: Materials Chemistry, Building Blocks for Small Molecule Semiconductors, Diphenylamines (for High-Performance Polymer Research)
Other Product Names / Synonyms: Di-p-anisylamine, Bis(4-methoxyphenyl)amine
Formula:
C₁₄H₁₅NO₂ MW: 229.28 g/mol Melting Pt: 103 °C |
MDL Number:
MFCD00014895 CAS Number: 101-70-2 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...