Trisodium 2-(4-Sulfophenylazo)-1,8-dihydroxynaphthalene-3,6-disulfonate (SPADNS) ≥95.0% (by HPLC)
Supplier: TCI America
D0604-5G
D0604-25G
TCD0604-005GEA
135.88
CAD
TCD0604-005G
TCD0604-025G
Trisodium 2-(4-Sulfophenylazo)-1,8-dihydroxynaphthalene-3,6-disulfonate (SPADNS) ≥95.0% (by HPLC)
Trisodium 2-(4-Sulfophenylazo)-1,8-dihydroxynaphthalene-3,6-disulfonate (SPADNS)
CAS Number: 23647-14-5
MDL Number: MFCD00003952
Molecular Formula: C16H12N2O11S3
Molecular Weight: 570.40
Purity/Analysis Method: >95.0% (HPLC)
Form: Crystal
Product Categories: Analytical Science, Azo Compounds (Chelating Agents)
Formula:
C₁₆H₁₂N₂O₁₁S₃ |
MDL Number:
MFCD00003952 CAS Number: 23647-14-5 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...