Ethyl red ≥98.0% (by titrimetric analysis)
Supplier: TCI America
D0524-25G
D0524-1G
TCD0524-025GEA
192.76
CAD
TCD0524-025G
TCD0524-001G
Ethyl red ≥98.0% (by titrimetric analysis)
Ethyl red
[not Cyanin dyes]
CAS Number: 76058-33-8
MDL Number: MFCD00002427
Molecular Formula: C17H19N3O2
Molecular Weight: 297.36
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 146
Other Product Names / Synonyms: 4-(Diethylamino)azobenzene-2'-carboxylic Acid, Diethyl Red
Formula:
C₁₇H₁₉N₃O₂ Melting Pt: 146 °C |
MDL Number:
MFCD00002427 CAS Number: 76058-33-8 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...