1,3-Dicyclohexyl-2-thiourea ≥98.0% (by total nitrogen basis)

Supplier: TCI America

D0440-25G D0440-5G
TCD0440-025GEA 67.94 CAD
TCD0440-025G 76218-956
1,3-Dicyclohexyl-2-thiourea ≥98.0% (by total nitrogen basis)
1,3-Dicyclohexyl-2-thiourea

CAS Number: 1212-29-9
MDL Number: MFCD00021316
Molecular Formula: C13H24N2S
Molecular Weight: 240.41
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 181

Formula: C₁₃H₂₄N₂S
Melting Pt: 181 °C
MDL Number: MFCD00021316
CAS Number: 1212-29-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5a354339601843e726fa7bebc81cd7398892d1af&vid=5a354339601843e726fa7bebc81cd7398892d1af&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (N)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR